siRNA Details
Gene Symbol: C1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106021.1 | NP_001099491.1 | 5 | 1012 | ||
XM_006251516.2 | XP_006251578.1 | 6 | 1131 | |||
XM_008770422.1 | XP_008768644.1 | 5 | 3194 | |||
XM_008770423.1 | XP_008768645.1 | 6 | 3093 | |||
GenBank mRNA | BC168684.1 | - | 5 | 988 | 7930158 | |
FQ229879.1 | - | 5 | 941 | |||
FQ229896.1 | - | 5 | 990 |
