siRNA Details
Gene Symbol: C1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106021.1 | NP_001099491.1 | 5 | 1019 | ||
XM_006251516.2 | XP_006251578.1 | 6 | 1138 | |||
XM_008770422.1 | XP_008768644.1 | 5 | 3201 | |||
XM_008770423.1 | XP_008768645.1 | 6 | 3100 | |||
GenBank mRNA | BC168684.1 | - | 5 | 995 | 7930158 | |
FQ229879.1 | - | 5 | 948 | |||
FQ229896.1 | - | 5 | 997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
