siRNA Details
Gene Symbol: Fam161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013876.2 | NP_001013898.2 | 3 | 1396 | ||
XM_006251574.2 | XP_006251636.2 | 3 | 1396 | |||
XM_008770425.1 | XP_008768647.1 | 3 | 1396 | |||
GenBank mRNA | BC079233.1 | - | 3 | 1290 | 7132584 |
