siRNA Details
Gene Symbol: Erlec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106023.1 | NP_001099493.2 | 4 | 448 | ||
XM_006251615.2 | XP_006251677.1 | 4 | 766 | |||
GenBank mRNA | FQ228963.1 | - | 3 | 188 | ||
FQ232613.1 | - | 3 | 187 |
