siRNA Details
Gene Symbol: Tmed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007619.1 | NP_001007620.1 | 2 | 333 | ||
GenBank mRNA | AY539871.1 | - | 245 | |||
BC079452.1 | - | 2 | 333 | 7121068 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028876, AK159813, BC132023, AK151650, BC138226, and others.
Gene Ontology Categories:
