siRNA Details
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007620.1 | NP_001007621.1 | 9 | 1153 | ||
XM_006251754.1 | XP_006251816.1 | 10 | 1013 | |||
GenBank mRNA | BC079137.1 | - | 9 | 1153 | 7127701 | |
FQ212778.1 | - | 9 | 841 | |||
FQ214416.1 | - | 9 | 842 | |||
FQ234205.1 | - | 9 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
