siRNA Details
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100509.1 | NP_001093979.1 | 9 | 665 | ||
GenBank mRNA | BC107950.1 | - | 9 | 586 | ||
FQ213949.1 | - | 9 | 613 | |||
FQ214043.1 | - | 9 | 618 | |||
FQ227742.1 | - | 9 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
