siRNA Details
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106029.2 | NP_001099499.1 | 2 | 139 | ||
GenBank mRNA | BC158747.1 | - | 2 | 139 | 9101224 | |
FQ222749.1 | - | 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
