siRNA Details
Gene Symbol: Nrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106036.2 | NP_001099506.1 | 1 | 57 | ||
XM_006251935.2 | XP_006251997.1 | 3 | 336 | |||
XM_006251937.2 | XP_006251999.1 | 3 | 316 | |||
XM_008770672.1 | XP_008768894.1 | 2 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
