siRNA Details
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106039.1 | NP_001099509.1 | 2, 3 | 219 | ||
XM_008770675.1 | XP_008768897.1 | 2, 3 | 1347 |

More Information
Other Species Targeted

Gene Ontology Categories:
