siRNA Details
Gene Symbol: Mcpt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170466.1 | NP_001163937.1 | 4 | 521 | ||
GenBank mRNA | FQ219924.1 | - | 3 | 492 | ||
U67888.1 | - | 4 | 498 |
Gene Symbol: LOC103690000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770718.1 | XP_008768940.1 | 500 |
