siRNA Details
Gene Symbol: Ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106041.1 | NP_001099511.1 | 3 | 372 | ||
XM_006252041.1 | XP_006252103.1 | 3 | 284 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106041.1 | NP_001099511.1 | 3 | 372 | ||
XM_006252041.1 | XP_006252103.1 | 3 | 284 |