siRNA Details
Gene Symbol: Sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033685.2 | NP_001028857.2 | 3 | 266 | ||
GenBank mRNA | BC104706.1 | - | 3 | 255 | 7316605 | |
CK357541.1 | - | 3 | 223 | |||
FQ212380.1 | - | 3 | 238 | |||
FQ223271.1 | - | 3 | 233 | |||
FQ234134.1 | - | 3 | 274 |
Gene Symbol: RGD1561590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727811.4 | XP_002727857.1 | 1 | 289 |

More Information
Other Species Targeted

Gene Ontology Categories:
