siRNA Details
Gene Symbol: Dleu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106043.1 | NP_001099513.1 | 3 | 848 | ||
XM_006252176.2 | XP_006252238.1 | 2 | 1039 | |||
GenBank mRNA | BC158600.1 | - | 2 | 556 | 7130710 |

More Information
Other Species Targeted

