siRNA Details
Gene Symbol: Loxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106047.2 | NP_001099517.2 | 3, 4 | 871 | ||
GenBank mRNA | BC168900.1 | - | 3, 4 | 871 | 9032601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
