siRNA Details
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007622.1 | NP_001007623.1 | 10 | 1092 | ||
XM_006252255.2 | XP_006252317.1 | 10 | 1107 | |||
XM_006252256.2 | XP_006252318.1 | 10 | 1102 | |||
XM_006252257.2 | XP_006252319.1 | 10 | 1624 | |||
XM_006252258.1 | XP_006252320.1 | 10 | 992 | |||
XM_006252259.2 | XP_006252321.1 | 10 | 1659 | |||
XM_008770823.1 | XP_008769045.1 | 10 | 993 | |||
GenBank mRNA | AY531526.1 | - | 10 | 1074 | ||
BC079091.1 | - | 10 | 1092 | 7123600 |
