siRNA Details
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007622.1 | NP_001007623.1 | 2, 3 | 204 | ||
XM_006252255.2 | XP_006252317.1 | 2, 3 | 222 | |||
XM_006252256.2 | XP_006252318.1 | 2, 3 | 214 | |||
XM_006252257.2 | XP_006252319.1 | 2, 3 | 736 | |||
XM_006252258.1 | XP_006252320.1 | 2, 3 | 104 | |||
XM_006252259.2 | XP_006252321.1 | 2, 3 | 771 | |||
XM_008770823.1 | XP_008769045.1 | 2, 3 | 105 | |||
GenBank mRNA | AY531526.1 | - | 2, 3 | 186 | ||
BC079091.1 | - | 2, 3 | 204 | 7123600 |
