siRNA Details
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006963.1 | NP_001006964.1 | 4 | 631 | ||
GenBank mRNA | BC083735.1 | - | 4 | 631 | 7191589 | |
FQ210227.1 | - | 4 | 661 | |||
FQ212925.1 | - | 4 | 588 | |||
FQ218718.1 | - | 4 | 630 | |||
FQ229426.1 | - | 4 | 630 | |||
FQ230065.1 | - | 4 | 603 |

More Information
Other Species Targeted

Gene Ontology Categories:
