siRNA Details
Gene Symbol: Nudt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106049.1 | NP_001099519.1 | 3 | 626 | ||
XM_006252294.2 | XP_006252356.1 | 2 | 941 | |||
XM_008770862.1 | XP_008769084.1 | 3 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
