siRNA Details
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013051.1 | NP_001013069.1 | 4 | 251 | ||
GenBank mRNA | BC158724.1 | - | 4 | 251 | 9090831 | |
FQ220550.1 | - | 4 | 286 | |||
FQ221379.1 | - | 4 | 277 | |||
FQ225182.1 | - | 4 | 369 | |||
FQ227975.1 | - | 4 | 369 | |||
FQ232174.1 | - | 4 | 369 |

More Information
Other Species Targeted

