siRNA Details
Gene Symbol: RGD1306441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106053.1 | NP_001099523.1 | 2 | 955 | ||
XM_006252385.2 | XP_006252447.1 | 2 | 952 | |||
XM_006252386.1 | XP_006252448.1 | 3 | 1322 | |||
GenBank mRNA | BC079439.1 | - | 3 | 1327 |
