siRNA Details
Gene Symbol: Fam213a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008297.1 | NP_001008298.1 | 3, 4 | 471 | ||
XM_006252573.1 | XP_006252635.1 | 3, 4 | 419 | |||
GenBank mRNA | BC086323.1 | - | 3, 4 | 471 | 7304524 | |
FQ213064.1 | - | 305 | ||||
FQ220194.1 | - | 3, 4 | 298 |
