siRNA Details
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106061.1 | NP_001099531.1 | 8 | 1050 | ||
XM_006252574.2 | XP_006252636.1 | 11 | 1182 | |||
XM_006252575.2 | XP_006252637.1 | 11 | 1380 | |||
XM_006252576.2 | XP_006252638.1 | 9 | 1546 | |||
XM_008770982.1 | XP_008769204.1 | 11 | 1247 | |||
XM_008770983.1 | XP_008769205.1 | 11 | 1089 |
