siRNA Details
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252574.2 | XP_006252636.1 | 4 | 333 | ||
XM_006252575.2 | XP_006252637.1 | 4 | 531 | |||
XM_006252576.2 | XP_006252638.1 | 2 | 697 | |||
XM_008770982.1 | XP_008769204.1 | 4 | 398 | |||
XM_008770983.1 | XP_008769205.1 | 4 | 240 |
