siRNA Details
Gene Symbol: Selk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207589.3 | NP_997472.2 | 2 | 148 | ||
GenBank mRNA | AY319924.1 | - | 2 | 71 | ||
BC088086.1 | - | 2 | 92 | 7326199 | ||
CF979418.1 | - | 2 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
