siRNA Details
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198731.2 | NP_942026.1 | 2 | 777 | ||
XM_006252578.2 | XP_006252640.1 | 2 | 842 | |||
XM_006252579.2 | XP_006252641.1 | 2 | 796 | |||
XM_006252580.2 | XP_006252642.1 | 2 | 602 | |||
XM_008770984.1 | XP_008769206.1 | 2 | 1145 | |||
XM_008770985.1 | XP_008769207.1 | 3 | 1234 | |||
GenBank mRNA | AY365023.1 | - | 1 | 526 | ||
BC085787.1 | - | 2 | 777 | 7108667 | ||
X94769.1 | - | 1 | 52 |
