siRNA Details
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135866.1 | NP_001129338.1 | 12 | 1302 | ||
XM_006252581.2 | XP_006252643.1 | 12 | 1313 |

More Information
Other Species Targeted

Gene Ontology Categories:
