siRNA Details
Gene Symbol: Tnnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034105.1 | NP_001029277.1 | 5 | 392 | ||
XM_008770987.1 | XP_008769209.1 | 6 | 536 | |||
GenBank mRNA | DQ062205.1 | - | 5 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
