siRNA Details
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007623.1 | NP_001007624.1 | 5 | 797 | ||
XM_006252582.2 | XP_006252644.1 | 5 | 782 | |||
XM_006252583.2 | XP_006252645.1 | 5 | 785 | |||
XM_008770988.1 | XP_008769210.1 | 5 | 782 | |||
GenBank mRNA | BC079309.1 | - | 5 | 797 | 7134747 | |
FQ211422.1 | - | 5 | 806 |
