siRNA Details
Gene Symbol: LOC103693904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771162.1 | XP_008769384.1 | 2 | 186 |
Gene Symbol: RGD1564225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252782.2 | XP_006252844.1 | 2 | 197 |
