siRNA Details
Gene Symbol: LOC290595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106063.2 | NP_001099533.2 | 3 | 392 | ||
GenBank mRNA | AF152002.1 | - | 3 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
