siRNA Details
Gene Symbol: RGD1311847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013879.1 | NP_001013901.1 | 2 | 148 | ||
XM_006252799.2 | XP_006252861.1 | 2 | 205 | |||
XM_008771073.1 | XP_008769295.1 | 2 | 198 | |||
GenBank mRNA | BC079430.1 | - | 2 | 148 | 7114707 |
