siRNA Details
Gene Symbol: Cyp4f18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033686.1 | NP_001028858.1 | 709 | |||
GenBank mRNA | BC101918.1 | - | 709 | 7935301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
