siRNA Details
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106069.1 | NP_001099539.1 | 8 | 666 | ||
XM_006252864.2 | XP_006252926.1 | 8 | 1067 | |||
XM_006252865.2 | XP_006252927.1 | 8 | 1071 | |||
XM_006252866.2 | XP_006252928.2 | 7 | 645 | |||
XM_008771082.1 | XP_008769304.1 | 7 | 645 | |||
XM_008771083.1 | XP_008769305.1 | 8 | 721 | |||
XM_008771084.1 | XP_008769306.1 | 7 | 645 |
