siRNA Details
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252864.2 | XP_006252926.1 | 14, 15 | 1486 | ||
XM_006252865.2 | XP_006252927.1 | 14, 15 | 1490 | |||
XM_006252866.2 | XP_006252928.2 | 13, 14 | 1064 | |||
XM_008771082.1 | XP_008769304.1 | 13, 14 | 1064 | |||
XM_008771083.1 | XP_008769305.1 | 14, 15 | 1140 | |||
XM_008771084.1 | XP_008769306.1 | 13, 14 | 1064 |
