siRNA Details
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106069.1 | NP_001099539.1 | 7, 8 | 593 | ||
XM_006252864.2 | XP_006252926.1 | 7, 8 | 994 | |||
XM_006252865.2 | XP_006252927.1 | 7, 8 | 998 | |||
XM_006252866.2 | XP_006252928.2 | 6, 7 | 572 | |||
XM_008771082.1 | XP_008769304.1 | 6, 7 | 572 | |||
XM_008771083.1 | XP_008769305.1 | 7, 8 | 648 | |||
XM_008771084.1 | XP_008769306.1 | 6, 7 | 572 |

More Information
Other Species Targeted

Gene Ontology Categories:
