siRNA Details
Gene Symbol: Mpv17l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106072.2 | NP_001099542.1 | 4 | 560 | ||
XM_008771087.1 | XP_008769309.1 | 4 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
