siRNA Details
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201988.2 | NP_973717.1 | 5 | 480 | ||
GenBank mRNA | AB098134.1 | - | 6 | 475 | ||
BC098645.1 | - | 5 | 480 | 7108516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
