siRNA Details
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037180.1 | NP_001032257.1 | 3 | 353 | ||
GenBank mRNA | BC107454.1 | - | 3 | 353 | 7378812 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037180.1 | NP_001032257.1 | 3 | 353 | ||
GenBank mRNA | BC107454.1 | - | 3 | 353 | 7378812 |