siRNA Details
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106074.1 | NP_001099544.1 | 7 | 1270 | ||
XM_006252872.2 | XP_006252934.1 | 7 | 1313 | |||
XM_006252874.2 | XP_006252936.1 | 7 | 1313 |
