siRNA Details
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013881.2 | NP_001013903.2 | 10 | 1878 | ||
XM_006252879.2 | XP_006252941.1 | 9 | 1558 | |||
XM_006252880.2 | XP_006252942.1 | 9 | 1555 | |||
XM_006252881.2 | XP_006252943.1 | 9 | 1549 | |||
XM_006252882.2 | XP_006252944.1 | 9 | 1546 | |||
XM_006252883.2 | XP_006252945.1 | 9 | 1558 | |||
XM_006252884.2 | XP_006252946.1 | 10 | 1837 | |||
XM_006252887.2 | XP_006252949.1 | 9 | 1370 | |||
XM_006252889.2 | XP_006252951.1 | 9 | 1556 | |||
XM_008771089.1 | XP_008769311.1 | 11 | 1927 | |||
XM_008771090.1 | XP_008769312.1 | 10 | 1557 | |||
GenBank mRNA | BC089902.1 | - | 7 | 968 | 7383529 |
