siRNA Details
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013882.2 | NP_001013904.2 | 3 | 440 | ||
NM_001161512.1 | NP_001154984.1 | 3 | 656 | |||
GenBank mRNA | BC087138.1 | - | 3 | 395 | 7125992 | |
BP504069.1 | - | 658 | ||||
DY317307.1 | - | 3 | 408 | |||
EV776012.1 | - | 3 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
