siRNA Details
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047086.1 | NP_001040551.1 | 4 | 452 | ||
XM_006253139.2 | XP_006253201.1 | 4 | 713 | |||
XM_008771256.1 | XP_008769478.1 | 4 | 548 | |||
GenBank mRNA | BC166593.1 | - | 4 | 493 | 7934031 | |
DQ674266.1 | - | 4 | 452 |
