siRNA Details
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017443.1 | NP_001017443.1 | 7 | 943 | ||
XM_006253141.2 | XP_006253203.1 | 7 | 859 | |||
XM_006253143.2 | XP_006253205.1 | 8 | 1940 | |||
XM_008771257.1 | XP_008769479.1 | 6 | 745 | |||
GenBank mRNA | BC092575.1 | - | 7 | 943 | 7114719 |
