siRNA Details
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106084.2 | NP_001099554.2 | 25 | 3634 | ||
XM_008771295.1 | XP_008769517.1 | 25 | 3682 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106084.2 | NP_001099554.2 | 25 | 3634 | ||
XM_008771295.1 | XP_008769517.1 | 25 | 3682 |