siRNA Details
Gene Symbol: RGD1564788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253254.2 | XP_006253316.1 | 32 | 4525 | ||
XM_006253256.2 | XP_006253318.1 | 32 | 3954 | |||
XM_008771311.1 | XP_008769533.1 | 30 | 4525 |
