siRNA Details
Gene Symbol: Rpl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173340.2 | NP_775462.2 | 3, 4 | 168 | ||
GenBank mRNA | BC086382.1 | - | 3, 4 | 168 | 7301486 | |
EU000472.1 | - | 3, 4 | 168 | |||
X68282.1 | - | 3, 4 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
