siRNA Details
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106092.2 | NP_001099562.2 | 8 | 626 | ||
GenBank mRNA | BC160860.1 | - | 8 | 626 | 7321877 | |
FQ217181.1 | - | 8 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
