siRNA Details
Gene Symbol: Tmem255b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771387.1 | XP_008769609.1 | 4, 5 | 552 | ||
XM_008771388.1 | XP_008769610.1 | 3, 4 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
