siRNA Details
Gene Symbol: Adprhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013054.1 | NP_001013072.1 | 5 | 700 | ||
XM_006253439.2 | XP_006253501.1 | 5 | 1174 | |||
GenBank mRNA | BC083773.1 | - | 5 | 700 | 7193406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
